1,6-Anhydro-β-D-glucopyranose, 99+%
1,6-Anhydro-β-D-glucopyranose, 99+%
1,6-Anhydro-β-D-glucopyranose, 99+%
1,6-Anhydro-β-D-glucopyranose, 99+%
Thermo Scientific Chemicals
1,6-Anhydro-β-D-glucopyranose, 99+%
CAS: 498-07-7 | C6H10O5 | 162.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Packaging:
Glass bottle
Catalog number ACR270950010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS107-98-2
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Color scale=<10 APHA
Infrared spectrumConforms
Refractive index1.4020 to 1.4040 (20°C, 589 nm)
Acidity (CH3COOH)=<0.01 %