PYBOP(R), 99%, Thermo Scientific Chemicals
PYBOP(R), 99%, Thermo Scientific Chemicals
PYBOP(R), 99%, Thermo Scientific Chemicals
PYBOP(R), 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
PYBOP(R), 99%, Thermo Scientific Chemicals
CAS: 128625-52-5 | C18H28F6N6OP2 | 520.39 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Catalog number ACR270720050
Price (MYR)
689.00
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
689.00
Add to cart
Chemical Identifiers
CAS7785-26-4
IUPAC Name(1S,5S)-2,6,6-trimethylbicyclo[3.1.1]hept-2-ene
Molecular FormulaC10H16
InChI KeyGRWFGVWFFZKLTI-IUCAKERBSA-N
SMILESCC1=CC[C@H]2C[C@@H]1C2(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Refractive index1.4640 to 1.4680 (20°C, 589 nm)
Specific gravity(20°C) 0.858 to 0.861
Color scale=<30 APHA
Infrared spectrumConforms
View more
Coupling reagent
RUO – Research Use Only