Alginic acid
Alginic acid
Alginic acid
Thermo Scientific Chemicals
Alginic acid
CAS: 9005-32-7 | (C6H8O7)A(C6H8O7)B
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Plastic bottle
Catalog number ACR270585000
Price (MYR)
2,014.00
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
2,014.00
Add to cart
Chemical Identifiers
CAS1310-73-2
IUPAC Namesodium hydroxide
Molecular FormulaHNaO
InChI KeyHEMHJVSKTPXQMS-UHFFFAOYSA-M
SMILES[OH-].[Na+]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Suppliers CofA2.0N ± 0.24%
Appearance (Color)Clear colorless