Alginic acid
Alginic acid
Alginic acid
Thermo Scientific Chemicals
Alginic acid
CAS: 9005-32-7 | (C6H8O7)A(C6H8O7)B
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Catalog number ACR270581000
Price (MYR)
612.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
612.00
Add to cart
Chemical Identifiers
CAS121-79-9
IUPAC Namepropyl 3,4,5-trihydroxybenzoate
Molecular FormulaC10H12O5
InChI KeyZTHYODDOHIVTJV-UHFFFAOYSA-N
SMILESCCCOC(=O)C1=CC(O)=C(O)C(O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light beige
Loss on drying=<0.5 % (110°C, 4 hrs)
Heavy metals (as Pb)=<10 ppm
Sulfated ash=<0.1 %
Appearance (Form)Crystalline powder