1,4-Dioxane, 99.5%, for analysis, stabilized, Thermo Scientific Chemicals
1,4-Dioxane, 99.5%, for analysis, stabilized, Thermo Scientific Chemicals
1,4-Dioxane, 99.5%, for analysis, stabilized, Thermo Scientific Chemicals
1,4-Dioxane, 99.5%, for analysis, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,4-Dioxane, 99.5%, for analysis, stabilized, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Packaging:
Glass Bottle
Catalog number ACR270490010
Price (MYR)
625.00
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
625.00
Add to cart
Chemical Identifiers
CAS9010-66-6
MDL NumberMFCD00062414
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to straw yellow to tan
Infrared spectrumConforms
Loss on drying=<8.0 % (105°C, 2 h)
Appearance (Form)Powder
Residue after ignition=<2.0 % (800°C)