Formic acid, 99%, for analysis, Thermo Scientific Chemicals
Formic acid, 99%, for analysis, Thermo Scientific Chemicals
Formic acid, 99%, for analysis, Thermo Scientific Chemicals
Formic acid, 99%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Formic acid, 99%, for analysis, Thermo Scientific Chemicals
Formic acid, 99%, CH2O2, CAS Number-64-18-6, acide formique, aminic acid, bilorin, collo-bueglatt, formira, formisoton, formylic acid, hydrogen carboxylic acid, methanoic acid, myrmicyl, 2.5L, Vapor Pressure: 44mbar at 20 deg.C, 02, 8, 101.0 deg.C, 0.1ppm max., CHEBI:30751, 5ppm max., Undesignated
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
25 mL
2.5 L
Packaging:
Plastic Bottle
Catalog number ACR270480010
Price (MYR)
1,006.00
Add to cart
Quantity:
1 L
Packaging:
Plastic Bottle
Request bulk or custom format
Price (MYR)
1,006.00
Add to cart
Chemical Identifiers
CAS7440-48-4
IUPAC Namecobalt
Molecular FormulaCo
InChI KeyGUTLYIVDDKVIGB-UHFFFAOYSA-N
SMILES[Co]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Blue silver
FormRod, 12.5mm (0.492in) diameter.
Assay from Supplier's CofA≥99.95% (metals basis)
For analysis
RUO – Research Use Only