Perfluorooctyl iodide, 98%, Thermo Scientific Chemicals
Perfluorooctyl iodide, 98%, Thermo Scientific Chemicals
Perfluorooctyl iodide, 98%, Thermo Scientific Chemicals
Perfluorooctyl iodide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Perfluorooctyl iodide, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
Packaging:
Glass bottle
Catalog number ACR269960100
Price (MYR)
707.00
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
707.00
Add to cart
Chemical Identifiers
CAS1718-34-9
IUPAC Namesodium (3Z)-3-[2-(4-nitrophenyl)hydrazin-1-ylidene]-6-oxocyclohexa-1,4-diene-1-carboxylate
Molecular FormulaC13H8N3NaO5
InChI KeyHKSANXZSKDUDBD-SOCRLDLMSA-M
SMILES[Na+].[O-]C(=O)C1=CC(C=CC1=O)=N/NC1=CC=C(C=C1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Orange to brown
Appearance (Form)Crystalline powder
Infrared spectrumConforms
UV spectrophotometry>=95 %
Loss on drying=<5 % (105°C)