Monomethyl monopotassium malonate, 98%, Thermo Scientific Chemicals
Monomethyl monopotassium malonate, 98%, Thermo Scientific Chemicals
Monomethyl monopotassium malonate, 98%, Thermo Scientific Chemicals
Monomethyl monopotassium malonate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Monomethyl monopotassium malonate, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number ACR269550250
Price (MYR)
464.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
464.00
Add to cart
Specifications
Chemical Name or MaterialMonomethyl monopotassium malonate
CAS38330-80-2
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0