Lithium acetate, anhydrous, 99%, extra pure, Thermo Scientific Chemicals
Lithium acetate, anhydrous, 99%, extra pure, Thermo Scientific Chemicals
Lithium acetate, anhydrous, 99%, extra pure, Thermo Scientific Chemicals
Lithium acetate, anhydrous, 99%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Lithium acetate, anhydrous, 99%, extra pure, Thermo Scientific Chemicals
CAS: 546-89-4 | C2H3LiO2 | 65.98 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
Catalog number ACR268640010
Price (MYR)
2,610.00
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (MYR)
2,610.00
Add to cart
Chemical Identifiers
CAS1576-35-8
IUPAC Name4-methylbenzene-1-sulfonohydrazide
Molecular FormulaC7H10N2O2S
InChI KeyICGLPKIVTVWCFT-UHFFFAOYSA-N
SMILESCC1=CC=C(C=C1)S(=O)(=O)NN
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
HPLC>=96.0 %
Appearance (Color)White to off-white to light grey
Infrared spectrumConforms
Melting point105°C to 113°C (Dec)