Ethyl acetate, +99.5%, for HPLC, Thermo Scientific Chemicals
Ethyl acetate, +99.5%, for HPLC, Thermo Scientific Chemicals
Ethyl acetate, +99.5%, for HPLC, Thermo Scientific Chemicals
Ethyl acetate, +99.5%, for HPLC, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ethyl acetate, +99.5%, for HPLC, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
Catalog number ACR268350025
Price (MYR)
749.00
Add to cart
Quantity:
2.5 L
Request bulk or custom format
Price (MYR)
749.00
Add to cart
Chemical Identifiers
CAS75-65-0
IUPAC Name2-methylpropan-2-ol
Molecular FormulaC4H10O
InChI KeyDKGAVHZHDRPRBM-UHFFFAOYSA-N
SMILESCC(C)(C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Boron (B)=<0.02 ppm
Magnesium (Mg)=<0.1 ppm
Nickel (Ni)=<0.02 ppm
Appearance (Color)Clear colorless
View more
For HPLC
RUO – Research Use Only