1,4-Dioxane, 99.5%, for HPLC, Thermo Scientific Chemicals
1,4-Dioxane, 99.5%, for HPLC, Thermo Scientific Chemicals
1,4-Dioxane, 99.5%, for HPLC, Thermo Scientific Chemicals
1,4-Dioxane, 99.5%, for HPLC, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,4-Dioxane, 99.5%, for HPLC, Thermo Scientific Chemicals
CAS: 123-91-1 | C4H8O2 | 88.106 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass Bottle
Catalog number ACR268340025
Price (MYR)
3,786.00
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
3,786.00
Add to cart
Specifications
Chemical Name or Material1, 4-Dioxane
Melting Point12.0°C
Absorbance(1 cm cell vs HPLC−grade water), (1 cm cell vs HPLC−grade water) at 0nm, 0.03 max. at 270nm, 0.05 max. at 260nm, 0.075 max. at 250nm, 0.125 max. at 240nm, 0.22 max. at 230nm, 0.4 max. at 220nm, 1 max. at 218nm
CAS123-91-1
CAS Min %99.4
View more
For HPLC
RUO – Research Use Only