Acetonitrile, 99.9%, for HPLC, far UV, conform Eur.Ph., Thermo Scientific Chemicals
Acetonitrile, 99.9%, for HPLC, far UV, conform Eur.Ph., Thermo Scientific Chemicals
Acetonitrile, 99.9%, for HPLC, far UV, conform Eur.Ph., Thermo Scientific Chemicals
Acetonitrile, 99.9%, for HPLC, far UV, conform Eur.Ph., Thermo Scientific Chemicals
Thermo Scientific Chemicals
Acetonitrile, 99.9%, for HPLC, far UV, conform Eur.Ph., Thermo Scientific Chemicals
CAS: 75-05-8 | C2H3N | 41.053 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
Packaging:
Glass Bottle
Catalog number ACR268260025
Price (MYR)
1,893.00
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
1,893.00
Add to cart
Chemical Identifiers
CAS28439-86-3
IUPAC Name1-butoxy-4-isocyanatobenzene
Molecular FormulaC11H13NO2
InChI KeyASFIDVOMDQBCNP-UHFFFAOYSA-N
SMILESCCCCOC1=CC=C(C=C1)N=C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to white to yellow
FormLiquid
Refractive Index1.5200-1.5240 @ 20?C
Assay (GC)≥97.5%
For HPLC
RUO – Research Use Only