2,2,4-Trimethylpentane, 99.5%, for analysis, Thermo Scientific Chemicals
Have Questions?
Change view
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
Catalog number ACR265440010
Price (MYR)
585.00
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
585.00
Add to cart
Specifications
Chemical Name or Material2, 2, 4-Trimethylpentane
Melting Point-107.0°C
CAS540-84-1
CAS Min %99.4
Health Hazard 1GHS Signal Word: Danger