Tris-phosphate, 98%
Tris-phosphate, 98%
Tris-phosphate, 98%
Thermo Scientific Chemicals
Tris-phosphate, 98%
CAS: 6992-39-8 | C4H14NO7P | 219.13 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR264952500250 gGlass bottle
Catalog number ACR264952500
Price (MYR)
2,463.00
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,463.00
Add to cart
Specifications
Chemical Name or MaterialTris-phosphate
Name Note98%
CAS6992-39-8
Melting Point168.0°C to 172.0°C