Molecular sieves 4A, 10 to 18 mesh, Thermo Scientific Chemicals
Molecular sieves 4A, 10 to 18 mesh, Thermo Scientific Chemicals
Molecular sieves 4A, 10 to 18 mesh, Thermo Scientific Chemicals
Molecular sieves 4A, 10 to 18 mesh, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Molecular sieves 4A, 10 to 18 mesh, Thermo Scientific Chemicals
CAS: 70955-01-0 | C20H25FN2O8 | 440.42 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
Packaging:
Glass bottle
Catalog number ACR264755000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS7440-02-0
IUPAC Namenickel
Molecular FormulaNi
InChI KeyPXHVJJICTQNCMI-UHFFFAOYSA-N
SMILES[Ni]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥99.98% (metals basis)
FormSlug, 6.35mm (0.25in) diameter.
Appearance (Color)Silvery
Drying agent
RUO – Research Use Only