Butyric acid sodium salt, 98%, Thermo Scientific Chemicals
Butyric acid sodium salt, 98%, Thermo Scientific Chemicals
Butyric acid sodium salt, 98%, Thermo Scientific Chemicals
Butyric acid sodium salt, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Butyric acid sodium salt, 98%, Thermo Scientific Chemicals
CAS: 156-54-7 | C4H7NaO2 | 110.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number ACR263190250
Price (MYR)
434.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
434.00
Add to cart
Specifications
Chemical Name or MaterialButyric acid sodium salt
CAS156-54-7
Melting Point250.0°C to 253.0°C
ColorWhite
Health Hazard 2GHS H Statement: