Sodium hydroxide, extra pure, 50 wt% solution in water, Thermo Scientific Chemicals
Sodium hydroxide, extra pure, 50 wt% solution in water, Thermo Scientific Chemicals
Sodium hydroxide, extra pure, 50 wt% solution in water, Thermo Scientific Chemicals
Sodium hydroxide, extra pure, 50 wt% solution in water, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Sodium hydroxide, extra pure, 50 wt% solution in water, Thermo Scientific Chemicals
CAS: 1310-73-2 | HNaO | 39.997 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Plastic Bottle
Catalog number ACR259860025
Price (MYR)
761.00
Add to cart
Quantity:
2.5 L
Packaging:
Plastic Bottle
Request bulk or custom format
Price (MYR)
761.00
Add to cart
Chemical Identifiers
CAS7632-51-1
IUPAC Namevanadium(4+) tetrachloride
Molecular FormulaCl4V
InChI KeyJTJFQBNJBPPZRI-UHFFFAOYSA-J
SMILES[Cl-].[Cl-].[Cl-].[Cl-].[V+4]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Iron (Fe)=<10 ppm
Appearance (Color)Red-brown
Assay>=99.0 % (VCl4)
Vanadium(V) Oxide (V2O5)=<0.05 %