Glycerol formal, 99+%, mixture of isomers, stabilized, Thermo Scientific Chemicals
Glycerol formal, 99+%, mixture of isomers, stabilized, Thermo Scientific Chemicals
Glycerol formal, 99+%, mixture of isomers, stabilized, Thermo Scientific Chemicals
Glycerol formal, 99+%, mixture of isomers, stabilized, Thermo Scientific Chemicals
Glycerol formal, 99+%, mixture of isomers, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Glycerol formal, 99+%, mixture of isomers, stabilized, Thermo Scientific Chemicals
CAS: 4740-78-7 | C4H8O3 | 104.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
Packaging:
Glass bottle
Catalog number ACR259775000
Price (MYR)
1,039.00
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,039.00
Add to cart
Chemical Identifiers
CAS4740-78-7, 5464-28-8
Specifications
Chemical Name or MaterialGlycerol formal
Name Note99+%
CAS4740-78-7
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Causes serious eye irritation.
Suspected of damaging fertility or the unborn child.