Check Order Status
N-Methylhippuric acid, 99%, Thermo Scientific™
Element Search
Structure Search
Thermo Scientific Chemicals
N-Methylhippuric acid, 99%, Thermo Scientific™
Have Questions?
Change view
Quantity:
5 g
Packaging:
Glass bottle
Catalog number
ACR259760050
Price (MYR)
542.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
542.00
Add to cart
Specifications
Chemical Name or Material
N-Methylhippuric acid
Name Note
99%
CAS
2568-34-5
CAS Min %
98.5
Health Hazard 1
GHS Signal Word:
Warning