3,7-Dimethyl-7-hydroxyoctanal, 97%
3,7-Dimethyl-7-hydroxyoctanal, 97%
3,7-Dimethyl-7-hydroxyoctanal, 97%
3,7-Dimethyl-7-hydroxyoctanal, 97%
Thermo Scientific Chemicals
3,7-Dimethyl-7-hydroxyoctanal, 97%
CAS: 107-75-5 | C10H20O2 | 172.26 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR25968025025 mLGlass bottle
Catalog number ACR259680250
Price (MYR)
506.00
Add to cart
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
506.00
Add to cart
Chemical Identifiers
CAS124-30-1
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
FormCrystals or powder or crystalline powder or prills or fused solid or clear liquid as melt
Appearance (Color)White to cream to yellow