Butyl butyrate, 98%, Thermo Scientific Chemicals
Butyl butyrate, 98%, Thermo Scientific Chemicals
Butyl butyrate, 98%, Thermo Scientific Chemicals
Butyl butyrate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Butyl butyrate, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Packaging:
Glass bottle
Catalog number ACR259591000
Price (MYR)
346.00
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
346.00
Add to cart
Chemical Identifiers
CAS937-14-4
IUPAC Name3-chlorobenzene-1-carboperoxoic acid
Molecular FormulaC7H5ClO3
InChI KeyNHQDETIJWKXCTC-UHFFFAOYSA-N
SMILESOOC(=O)C1=CC=CC(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Moist powder
3-Chlorobenzoic acid=<12 %
HPLCdi-m-Chlorobenzoyl peroxide: < 0.1% Typical
HPLC=<0.2 % di-m-Chlorobenzoyl peroxide
Titration Iodometric70 to 75 %