γ-Undecalactone, 98%, Thermo Scientific Chemicals
γ-Undecalactone, 98%, Thermo Scientific Chemicals
γ-Undecalactone, 98%, Thermo Scientific Chemicals
γ-Undecalactone, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
γ-Undecalactone, 98%, Thermo Scientific Chemicals
CAS: 104-67-6 | C11H20O2 | 184.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
Packaging:
Glass bottle
Catalog number ACR259490250
Price (MYR)
424.00
Add to cart
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
424.00
Add to cart
Specifications
Chemical Name or Materialγ-Undecalactone
CAS104-67-6
Health Hazard 2GHS H Statement
Harmful to aquatic life with long lasting effects.
Health Hazard 3GHS P Statement
Avoid release to the environment.
Dispose of contents/container to an approved waste disposal plant.