L(+)-Ribose, 99%, Thermo Scientific Chemicals
L(+)-Ribose, 99%, Thermo Scientific Chemicals
L(+)-Ribose, 99%, Thermo Scientific Chemicals
L(+)-Ribose, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
L(+)-Ribose, 99%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Catalog number ACR257910050
Price (MYR)
703.00
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
703.00
Add to cart
Specifications
Chemical Name or MaterialL(+)-Ribose
CAS24259-59-4
Melting Point84.0°C to 86.0°C
EINECS Number246-110-4
Infrared SpectrumAuthentic