3-Cyclohexyl-1-propanol, 99%
3-Cyclohexyl-1-propanol, 99%
3-Cyclohexyl-1-propanol, 99%
3-Cyclohexyl-1-propanol, 99%
Thermo Scientific Chemicals
3-Cyclohexyl-1-propanol, 99%
CAS: 1124-63-6 | C9H18O | 142.24 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR2558800505 mLGlass bottle
Catalog number ACR255880050
Price (MYR)
1,209.00
Add to cart
Quantity:
5 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,209.00
Add to cart
Chemical Identifiers
CAS434-64-0
IUPAC Name1,2,3,4,5-pentafluoro-6-(trifluoromethyl)benzene
Molecular FormulaC7F8
InChI KeyUSPWUOFNOTUBAD-UHFFFAOYSA-N
SMILESFC1=C(F)C(F)=C(C(F)=C1F)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Identification (FTIR)Conforms
Appearance (Color)Clear colorless
Assay (GC)≥98.0%
Refractive Index1.3655-1.3695 @ 20?C