3-Chloroperoxybenzoic acid, 70-75%, balance 3-Chlorobenzoic acid and water, Thermo Scientific Chemicals
3-Chloroperoxybenzoic acid, 70-75%, balance 3-Chlorobenzoic acid and water, Thermo Scientific Chemicals
3-Chloroperoxybenzoic acid, 70-75%, balance 3-Chlorobenzoic acid and water, Thermo Scientific Chemicals
3-Chloroperoxybenzoic acid, 70-75%, balance 3-Chlorobenzoic acid and water, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3-Chloroperoxybenzoic acid, 70-75%, balance 3-Chlorobenzoic acid and water, Thermo Scientific Chemicals
3-Chloroperoxybenzoic acid, 70-75%, C7H5ClO3, CAS Number-937-14-4, 535-80-8, 7732-18-5
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
1 kg
Packaging:
View all
Plastic Bottle
Catalog number ACR255791000
Price (MYR)
957.00
Add to cart
Quantity:
100 g
Packaging:
Plastic Bottle
Request bulk or custom format
Price (MYR)
957.00
Add to cart
Chemical Identifiers
CAS937-14-4, 535-80-8, 7732-18-5
Specifications
Chemical Name or Material3-Chloroperoxybenzoic Acid
Name Note70 - 75%
CAS937-14-4
CAS Min %70.0
Health Hazard 1GHS Signal Word: Danger
View more
Oxidation reagent
RUO – Research Use Only