3-Chloroperoxybenzoic acid, 70-75%, balance 3-Chlorobenzoic acid and water, Thermo Scientific Chemicals
3-Chloroperoxybenzoic acid, 70-75%, balance 3-Chlorobenzoic acid and water, Thermo Scientific Chemicals
3-Chloroperoxybenzoic acid, 70-75%, balance 3-Chlorobenzoic acid and water, Thermo Scientific Chemicals
3-Chloroperoxybenzoic acid, 70-75%, balance 3-Chlorobenzoic acid and water, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3-Chloroperoxybenzoic acid, 70-75%, balance 3-Chlorobenzoic acid and water, Thermo Scientific Chemicals
3-Chloroperoxybenzoic acid, 70-75%, C7H5ClO3, CAS Number-937-14-4, 535-80-8, 7732-18-5
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
1 kg
Packaging:
View all
Plastic Bottle
Catalog number ACR255790250
Price (MYR)
560.00
Add to cart
Quantity:
25 g
Packaging:
Plastic Bottle
Request bulk or custom format
Price (MYR)
560.00
Add to cart
Chemical Identifiers
CAS37859-43-1
IUPAC Name2-(chloromethyl)-1,3-benzothiazole
Molecular FormulaC8H6ClNS
InChI KeySERUZNHRWBXDOX-UHFFFAOYSA-N
SMILESClCC1=NC2=CC=CC=C2S1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>94.0%
Oxidation reagent
RUO – Research Use Only