Ammonium hydroxide, extra pure, 25% solution in water, Thermo Scientific Chemicals
Ammonium hydroxide, extra pure, 25% solution in water, Thermo Scientific Chemicals
Ammonium hydroxide, extra pure, 25% solution in water, Thermo Scientific Chemicals
Ammonium hydroxide, extra pure, 25% solution in water, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ammonium hydroxide, extra pure, 25% solution in water, Thermo Scientific Chemicals
CAS: 1336-21-6 | H5NO | 35.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
Catalog number ACR255210010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS198-55-0
IUPAC Nameperylene
Molecular FormulaC20H12
InChI KeyCSHWQDPOILHKBI-UHFFFAOYSA-N
SMILESC1=CC2=C3C(C=CC=C3C3=C4C(C=CC=C24)=CC=C3)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥98.0%
Appearance (Color)Yellow to orange to brown or orange-brown
Melting Point (clear melt)272-280?C