4-Amino-6-chloro-1,3-benzenedisulfonamide, 98%, Thermo Scientific Chemicals
4-Amino-6-chloro-1,3-benzenedisulfonamide, 98%, Thermo Scientific Chemicals
4-Amino-6-chloro-1,3-benzenedisulfonamide, 98%, Thermo Scientific Chemicals
4-Amino-6-chloro-1,3-benzenedisulfonamide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Amino-6-chloro-1,3-benzenedisulfonamide, 98%, Thermo Scientific Chemicals
CAS: 121-30-2 | C6H8ClN3O4S2 | 285.73 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number ACR251850050
Price (MYR)
409.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
409.00
Add to cart
Chemical Identifiers
CAS3430-17-9
IUPAC Name2-bromo-3-methylpyridine
Molecular FormulaC6H6BrN
InChI KeyPZSISEFPCYMBDL-UHFFFAOYSA-N
SMILESCC1=CC=CN=C1Br
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (GC)≥96.0%
Appearance (Color)Clear colorless to yellow
Refractive Index1.5665-1.5705 @ 20°C