Sodium hyaluronate, 95%, Thermo Scientific Chemicals
Sodium hyaluronate, 95%, Thermo Scientific Chemicals
Sodium hyaluronate, 95%, Thermo Scientific Chemicals
Sodium hyaluronate, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Sodium hyaluronate, 95%, Thermo Scientific Chemicals
CAS: 9067-32-7 | (C14H20NO11Na)n | 417.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Packaging:
Glass bottle
Catalog number ACR251770250
Price (MYR)
6,242.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
6,242.00
Add to cart
Chemical Identifiers
CAS7647-17-8
IUPAC Namecaesium(1+) chloride
Molecular FormulaClCs
InChI KeyAIYUHDOJVYHVIT-UHFFFAOYSA-M
SMILES[Cl-].[Cs+]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to white
Appearance (Form)Crystalline powder or crystals
Purity>=99.94 % (metals basis)
UV(50 % in water) 1 cm cell
UVat 260 nm A: =<0.02