Sodium hyaluronate, 95%, Thermo Scientific Chemicals
Sodium hyaluronate, 95%, Thermo Scientific Chemicals
Sodium hyaluronate, 95%, Thermo Scientific Chemicals
Sodium hyaluronate, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Sodium hyaluronate, 95%, Thermo Scientific Chemicals
CAS: 9067-32-7 | (C14H20NO11Na)n | 417.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Packaging:
Glass bottle
Catalog number ACR251770050
Price (MYR)
1,861.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,861.00
Add to cart
Chemical Identifiers
CAS7727-15-3
IUPAC Namealuminium(3+) tribromide
Molecular FormulaAlBr3
InChI KeyPQLAYKMGZDUDLQ-UHFFFAOYSA-K
SMILES[Al+3].[Br-].[Br-].[Br-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or chunks
Titration Complexometric>=98.0 %
Chloride (Cl)=<2000 ppm
Sulfate (SO4)=<200 ppm
Appearance (Color)White to yellow to pink to grey