Sodium hyaluronate, 95%, Thermo Scientific Chemicals
Sodium hyaluronate, 95%, Thermo Scientific Chemicals
Sodium hyaluronate, 95%, Thermo Scientific Chemicals
Sodium hyaluronate, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Sodium hyaluronate, 95%, Thermo Scientific Chemicals
CAS: 9067-32-7 | (C14H20NO11Na)n | 417.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number ACR251770010
Price (MYR)
659.00
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
659.00
Add to cart
Chemical Identifiers
CAS91-58-7
IUPAC Name2-chloronaphthalene
Molecular FormulaC10H7Cl
InChI KeyCGYGETOMCSJHJU-UHFFFAOYSA-N
SMILESClC1=CC=C2C=CC=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Melting point57°C to 60°C
Appearance (Color)White to off-white
Infrared spectrumConforms
GC>=98.5 %