1-Amino-4-methylpiperazine, 99%, Thermo Scientific Chemicals
1-Amino-4-methylpiperazine, 99%, Thermo Scientific Chemicals
1-Amino-4-methylpiperazine, 99%, Thermo Scientific Chemicals
1-Amino-4-methylpiperazine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Amino-4-methylpiperazine, 99%, Thermo Scientific Chemicals
CAS: 6928-85-4 | C5H13N3 | 115.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number ACR251390250
Price (MYR)
670.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
670.00
Add to cart
Specifications
Chemical Name or Material1-Amino-4-methylpiperazine
Name Note99%
CAS6928-85-4
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning