L-Lactic acid, sodium salt, 60 wt% solution in water
L-Lactic acid, sodium salt, 60 wt% solution in water
L-Lactic acid, sodium salt, 60 wt% solution in water
L-Lactic acid, sodium salt, 60 wt% solution in water
Thermo Scientific Chemicals
L-Lactic acid, sodium salt, 60 wt% solution in water
CAS: 867-56-1 | C3H5NaO3 | 112.06 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR2503000101 LGlass bottle
Catalog number ACR250300010
Price (MYR)
681.00
Add to cart
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
681.00
Add to cart
Chemical Identifiers
CAS7789-00-6
IUPAC Namedipotassium dioxochromiumbis(olate)
Molecular FormulaCrK2O4
InChI KeyXMXNVYPJWBTAHN-UHFFFAOYSA-N
SMILES[K+].[K+].[O-][Cr]([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
pH8.6 to 9.8 (5% soln. at 25°C)
Insoluble matter=<0.005 %
Chloride (Cl)=<0.005 %
Sulfate (SO4)=<0.03 %
Calcium (Ca)=<0.005 %