Calcium L-lactate pentahydrate, 98%
Calcium L-lactate pentahydrate, 98%
Calcium L-lactate pentahydrate, 98%
Calcium L-lactate pentahydrate, 98%
Thermo Scientific Chemicals
Calcium L-lactate pentahydrate, 98%
CAS: 865717-20-0 | C6H10CaO6·5H2O | 308.29 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR2502000101 kgPlastic bottle
Catalog number ACR250200010
Price (MYR)
576.00
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
576.00
Add to cart
Specifications
Chemical Name or MaterialCalcium L-lactate pentahydrate
Melting Point240.0°C
Name Note98%
CAS865717-20-0