n-Butyl lactate, 99%, Thermo Scientific Chemicals
n-Butyl lactate, 99%, Thermo Scientific Chemicals
n-Butyl lactate, 99%, Thermo Scientific Chemicals
n-Butyl lactate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
n-Butyl lactate, 99%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
500 mL
Packaging:
Glass bottle
Catalog number ACR250180025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Materialn-Butyl lactate
Name Note99%
CAS138-22-7
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning