1-Iodododecane, 98%, stabilized, Thermo Scientific Chemicals
1-Iodododecane, 98%, stabilized, Thermo Scientific Chemicals
1-Iodododecane, 98%, stabilized, Thermo Scientific Chemicals
1-Iodododecane, 98%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Iodododecane, 98%, stabilized, Thermo Scientific Chemicals
CAS: 4292-19-7 | C12H25I | 296.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Packaging:
Glass bottle
Catalog number ACR250091000
Price (MYR)
1,244.00
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,244.00
Add to cart
Chemical Identifiers
CAS590-28-3
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)96.1% min