1-Chloro-2-iodobenzene, 99%, Thermo Scientific Chemicals
1-Chloro-2-iodobenzene, 99%, Thermo Scientific Chemicals
1-Chloro-2-iodobenzene, 99%, Thermo Scientific Chemicals
1-Chloro-2-iodobenzene, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Chloro-2-iodobenzene, 99%, Thermo Scientific Chemicals
CAS: 615-41-8 | C6H4ClI | 238.45 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
Packaging:
Glass bottle
Catalog number ACR249460500
Price (MYR)
1,424.00
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,424.00
Add to cart
Specifications
Chemical Name or Material1-Chloro-2-iodobenzene
Name Note99%
CAS615-41-8
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning