Ethyl potassium malonate, 98%, Thermo Scientific Chemicals
Ethyl potassium malonate, 98%, Thermo Scientific Chemicals
Ethyl potassium malonate, 98%, Thermo Scientific Chemicals
Ethyl potassium malonate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ethyl potassium malonate, 98%, Thermo Scientific Chemicals
CAS: 6148-64-7 | C5H7KO4 | 170.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Catalog number ACR248080250
Price (MYR)
432.00
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
432.00
Add to cart
Specifications
Chemical Name or MaterialEthyl potassium malonate
Name Note98%
CAS6148-64-7
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement: