(2S,3S)-(+)-2,3-Butanediol, 99%, Thermo Scientific Chemicals
(2S,3S)-(+)-2,3-Butanediol, 99%, Thermo Scientific Chemicals
(2S,3S)-(+)-2,3-Butanediol, 99%, Thermo Scientific Chemicals
(2S,3S)-(+)-2,3-Butanediol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
(2S,3S)-(+)-2,3-Butanediol, 99%, Thermo Scientific Chemicals
CAS: 19132-06-0 | C4H10O2 | 90.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 g
Catalog number ACR247730025
Price (MYR)
2,031.00
Add to cart
Quantity:
2.5 g
Request bulk or custom format
Price (MYR)
2,031.00
Add to cart
Chemical Identifiers
CAS7646-85-7
IUPAC Namezinc(2+) dichloride
Molecular FormulaCl2Zn
InChI KeyJIAARYAFYJHUJI-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Zn++]
View more
SpecificationsSpecification SheetSpecification Sheet
AppearanceMay develop some turbidity or precipitate
Appearance (Color)Colorless to light yellow to light blue-grey
Appearance (Form)Solution
Titration Argentometric0.9 to 1.1 M
Water=<0.2 % (Coulometric)
View more
Chiral derivatization reagent
RUO – Research Use Only