2-Amino-5-nitrobenzoic acid, 98%, Thermo Scientific Chemicals
2-Amino-5-nitrobenzoic acid, 98%, Thermo Scientific Chemicals
2-Amino-5-nitrobenzoic acid, 98%, Thermo Scientific Chemicals
2-Amino-5-nitrobenzoic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Amino-5-nitrobenzoic acid, 98%, Thermo Scientific Chemicals
CAS: 616-79-5 | C7H6N2O4 | 182.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number ACR245950050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material2-Amino-5-nitrobenzoic acid
Name Note98%
CAS616-79-5
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning