3-Thiophenecarboxylic acid, 99%, Thermo Scientific Chemicals
3-Thiophenecarboxylic acid, 99%, Thermo Scientific Chemicals
3-Thiophenecarboxylic acid, 99%, Thermo Scientific Chemicals
3-Thiophenecarboxylic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3-Thiophenecarboxylic acid, 99%, Thermo Scientific Chemicals
CAS: 88-13-1 | C5H4O2S | 128.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number ACR243110050
Price (MYR)
864.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
864.00
Add to cart
Chemical Identifiers
CAS3034-19-3
IUPAC Name2-(2-nitrophenyl)hydrazinium chloride
Molecular FormulaC6H8ClN3O2
InChI KeyXCUBVSAYUSFHNN-UHFFFAOYSA-N
SMILES[Cl-].[NH3+]NC1=CC=CC=C1[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Orange to orange-brown to dark red
HPLC>=96.0 %
Infrared spectrumConforms
Water30 to 35 %
Appearance (Form)Powder