3-Thiophenecarboxylic acid, 99%, Thermo Scientific Chemicals
3-Thiophenecarboxylic acid, 99%, Thermo Scientific Chemicals
3-Thiophenecarboxylic acid, 99%, Thermo Scientific Chemicals
3-Thiophenecarboxylic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3-Thiophenecarboxylic acid, 99%, Thermo Scientific Chemicals
CAS: 88-13-1 | C5H4O2S | 128.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number ACR243110010
Price (MYR)
429.00
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
429.00
Add to cart
Specifications
Chemical Name or Material3-Thiophenecarboxylic acid
Name Note99%
CAS88-13-1
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning