Phloroglucinol, 99+%, anhydrous, Thermo Scientific Chemicals
Phloroglucinol, 99+%, anhydrous, Thermo Scientific Chemicals
Phloroglucinol, 99+%, anhydrous, Thermo Scientific Chemicals
Phloroglucinol, 99+%, anhydrous, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phloroglucinol, 99+%, anhydrous, Thermo Scientific Chemicals
CAS: 108-73-6 | C6H6O3 | 126.1 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Packaging:
Glass bottle
Catalog number ACR241765000
Price (MYR)
2,565.00
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,565.00
Add to cart
Specifications
Chemical Name or MaterialPhloroglucinol
CAS108-73-6
Health Hazard 1Warning
Health Hazard 2GHS H Statement
Harmful to aquatic life with long lasting effects.
Health Hazard 3GHS P Statement
Avoid release to the environment.
Dispose of contents/container to an approved waste disposal plant.