Phloroglucinol, 99+%, anhydrous, Thermo Scientific Chemicals
Phloroglucinol, 99+%, anhydrous, Thermo Scientific Chemicals
Phloroglucinol, 99+%, anhydrous, Thermo Scientific Chemicals
Phloroglucinol, 99+%, anhydrous, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phloroglucinol, 99+%, anhydrous, Thermo Scientific Chemicals
CAS: 108-73-6 | C6H6O3 | 126.1 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number ACR241760250
Price (MYR)
501.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
501.00
Add to cart
Chemical Identifiers
CAS110-17-8
IUPAC Name(2E)-but-2-enedioic acid
Molecular FormulaC4H4O4
InChI KeyVZCYOOQTPOCHFL-OWOJBTEDSA-N
SMILESOC(=O)\C=C\C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5% (non-U.S. specification)
Assay (Silylated GC)≥98.5% (non-U.S. specification)
Assay from Suppliers CofA≥98.5% (U.S. specification)
CommentSpecification differs for U.S. and non-U.S. material where indicated