Silica gel, for column chromatography, 0.060-0.200mm, 40Å
Silica gel, for column chromatography, 0.060-0.200mm, 40Å
Silica gel, for column chromatography, 0.060-0.200mm, 40Å
Silica gel, for column chromatography, 0.060-0.200mm, 40Å
Silica gel, for column chromatography, 0.060-0.200mm, 40Å
Thermo Scientific Chemicals
Silica gel, for column chromatography, 0.060-0.200mm, 40Å
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
Catalog number ACR241660010
Price (MYR)
1,068.00
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (MYR)
1,068.00
Add to cart
Chemical Identifiers
CAS104504-45-2
SpecificationsSpecification SheetSpecification Sheet
FormPowder or crystals or crystalline powder
Appearance (Color)White to cream
Assay from Supplier's CofA≥94.0% (HPLC)
Proton NMRConforms to structure
For column chromatography
RUO – Research Use Only