Urea hydrogen peroxide, 1 g tablets, stabilized, contains 35 wt% H2O2, Thermo Scientific Chemicals
Urea hydrogen peroxide, 1 g tablets, stabilized, contains 35 wt% H2O2, Thermo Scientific Chemicals
Urea hydrogen peroxide, 1 g tablets, stabilized, contains 35 wt% H2O2, Thermo Scientific Chemicals
Urea hydrogen peroxide, 1 g tablets, stabilized, contains 35 wt% H2O2, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Urea hydrogen peroxide, 1 g tablets, stabilized, contains 35 wt% H2O2, Thermo Scientific Chemicals
CAS: 124-43-6 | CH6N2O3 | 94.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
Packaging:
Glass bottle
Catalog number ACR241020010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialUrea hydrogen peroxide
CAS124-43-6
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes severe skin burns and eye damage.
May intensify fire; oxidizer.
CAS7722-88-5
View more
Oxidation reagent
RUO – Research Use Only