Silica gel, for chromatography, 0.200-0.500mm, 60Å
Silica gel, for chromatography, 0.200-0.500mm, 60Å
Silica gel, for chromatography, 0.200-0.500mm, 60Å
Silica gel, for chromatography, 0.200-0.500mm, 60Å
Thermo Scientific Chemicals
Silica gel, for chromatography, 0.200-0.500mm, 60Å
Have Questions?
Change viewbuttonViewtableView
Packaging:
Plastic Bottle
Quantity:
1 kg
Catalog number ACR240380010
Price (MYR)
775.00
Add to cart
Packaging:
Plastic Bottle
Quantity:
1 kg
Request bulk or custom format
Price (MYR)
775.00
Add to cart
Chemical Identifiers
CAS402-49-3
IUPAC Name1-(bromomethyl)-4-(trifluoromethyl)benzene
Molecular FormulaC8H6BrF3
InChI KeyIKSNDOVDVVPSMA-UHFFFAOYSA-N
SMILESFC(F)(F)C1=CC=C(CBr)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=97.5 %
Appearance (Color)White to light yellow
Appearance (Form)Low melting crystalline solid
Infrared spectrumConforms
Melting point30°C to 34°C
Applications:
For column chromatography
RUO – Research Use Only