Silica Gel, for Column Chromatography, 0.060-0.200mm, 60Å
Silica Gel, for Column Chromatography, 0.060-0.200mm, 60Å
Silica Gel, for Column Chromatography, 0.060-0.200mm, 60Å
Silica Gel, for Column Chromatography, 0.060-0.200mm, 60Å
Silica Gel, for Column Chromatography, 0.060-0.200mm, 60Å
Thermo Scientific Chemicals
Silica Gel, for Column Chromatography, 0.060-0.200mm, 60Å
For column chromatography.
Have Questions?
Change viewbuttonViewtableView
Quantity:
30 kg
5 kg
Catalog number ACR240370300
Price (MYR)
12,636.00
Add to cart
Quantity:
30 kg
Request bulk or custom format
Price (MYR)
12,636.00
Add to cart
Chemical Identifiers
CAS588-93-2
IUPAC Name1-bromo-4-propylbenzene
Molecular FormulaC9H11Br
InChI KeyNUPWGLKBGVNSJX-UHFFFAOYSA-N
SMILESCCCC1=CC=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.5345-1.5375 @ 20?C
Appearance (Color)Clear colorless
Assay (GC)≥98.5%