Silica Gel, for Column Chromatography, 0.060-0.200mm, 60Å
Silica Gel, for Column Chromatography, 0.060-0.200mm, 60Å
Silica Gel, for Column Chromatography, 0.060-0.200mm, 60Å
Silica Gel, for Column Chromatography, 0.060-0.200mm, 60Å
Silica Gel, for Column Chromatography, 0.060-0.200mm, 60Å
Thermo Scientific Chemicals
Silica Gel, for Column Chromatography, 0.060-0.200mm, 60Å
For column chromatography.
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
30 kg
Catalog number ACR240370050
Price (MYR)
3,083.00
Add to cart
Quantity:
5 kg
Request bulk or custom format
Price (MYR)
3,083.00
Add to cart
Chemical Identifiers
CAS110-18-9
IUPAC Name[2-(dimethylamino)ethyl]dimethylamine
Molecular FormulaC6H16N2
InChI KeyKWYHDKDOAIKMQN-UHFFFAOYSA-N
SMILESCN(C)CCN(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4170 to 1.4190 (20°C, 589 nm)
GC>=99.4 %