Silica gel, for column chromatography, 0.035-0.070mm, 60Å
Silica gel, for column chromatography, 0.035-0.070mm, 60Å
Silica gel, for column chromatography, 0.035-0.070mm, 60Å
Silica gel, for column chromatography, 0.035-0.070mm, 60Å
Silica gel, for column chromatography, 0.035-0.070mm, 60Å
Thermo Scientific Chemicals
Silica gel, for column chromatography, 0.035-0.070mm, 60Å
For column chromatography.
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
1 kg
Catalog number ACR240360050
Price (MYR)
3,689.00
Add to cart
Quantity:
5 kg
Request bulk or custom format
Price (MYR)
3,689.00
Add to cart
Chemical Identifiers
CAS92-84-2
IUPAC Name10H-phenothiazine
Molecular FormulaC12H9NS
InChI KeyWJFKNYWRSNBZNX-UHFFFAOYSA-N
SMILESN1C2=CC=CC=C2SC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
Melting point>=184°C
Appearance (Form)Beads or prills
HPLC>=98.5 %
Infrared spectrumConforms
For column chromatography
RUO – Research Use Only